Learn More
Indofine Chemical D-(+)-Xylose, 98%, 100 Gm
SDP
Supplier:  Indofine Chemical 024312 100 GM
Encompass_Preferred
D-(+)-XYLOSE 100 GM, Rare Organics & Biochemicals, C5H10O5, MW: 150.14, 98+%, 58-86-6, MFCD00151475
Catalog No. 50-908-597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.