Learn More
Chondrex Inc D-Xylose Assay Kit
Supplier:  Chondrex Inc 6601
Encompass_Preferred
D-Xylose Assay Kit in a convenient 96-well plate format to assay the D-Xylose, a simple monosaccharide, forevaluate transcellular uptake. ASSAY TYPE: Fluorescence Assay, ASSAY TIME: 1 hour, STANDARD RANGE: 20 mM to 0.3 mM, NUMBER OF SAMPLES: Up to 39 (duplicate) samples/plate. SAMPLE TYPES: Serum and Plasma
Catalog No. NC1929624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.