Learn More
Eurofins Discoverx [D-Tyr6, β-Ala11, Phe13, Nle14]-Bombesin(6-14), 1 mg. (92-1158)
Supplier:  Eurofins Discoverx 921158
Encompass
[D - Tyr6, ß - Ala11, Phe13, Nle14] - Bombesin (6-14) is a 9 amino acid active fragment of the neuropeptide Bombesin, and is a potent agonist of the orphan bombesin receptor subtype 3 (hBRS-3). Also an agonist of the G-protein-coupled receptors BBR1, BBR2, and BBR3. Bombesin biological effects include hunger control, stimulation of gastrin release from G-cells, hormone release, stimulation of pancreatic enzyme secretion, inhibition of gastric emptying, and modulation of gastric acid secretion. Provided as 1 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9688
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.