Learn More
Montana Molecular Green Downward GENIe cGMP Sensor Kit D0800G, 10mL, CMV
SDP
Supplier:  Montana Molecular D0800G010
Encompass_Preferred
Measure cyclic GMP (cGMP) signaling dynamics and ascertain the specificity of PDE inhibitors or SNP activators with this live cell fluorescent assay. Add GENIe to your cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. Decreasing green fluorescence indicates cGMP increase. The assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. Kit includes soluble guanylate cyclase..
Catalog No. 50-163-1935
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.