Learn More
ATCC D1.1 T Lymphoblast Cell Line (Human), Suspension Culture, BSL 1
Supplier: ATCC CRL3600
A human T lymphoblast cell line grown in suspension, derived as a CD4-negative Jurkat progeny with contact-dependent helper activity. Intended for immunology and immune disorder research; handled at biosafety level 1.
- Derived from Jurkat cells; CD4 negative.
- Suspension-growing T lymphoblast; male origin.
- Recommended medium: RPMI-1640 with 10% fetal bovine serum.
- Culture conditions: 37°C, 95% air/5% CO2; maintain 2 x 10^5-1 x 10^6 viable cells/mL.
- Cryopreservation: use complete medium with 5% DMSO; store in liquid nitrogen vapor.
- Biosafety level 1; standard microbiological practices recommended.
- Suitable for immunology and immune system disorder research.