Learn More
Biocare Medical, LLC D2-40 (Lymphatic Marker) Antibody - Predilute, 6.0 ml
Supplier: Biocare Medical, LLC PM266AA
D2-40 antibody is a selective marker of lymphatic endothelium in normal tissues and vascular lesions. Studies have shown [D2-40] staining in lymphatic channel endothelium, but not in the adjacent capillary. In the same study, D2-40 stained endothelium of lymphangiomas; whereas hemangiomas, glomus tumors, angiolipomas, pyogenic granulomas and vascular malformations were negative for staining. D2-40 has also been shown to react with Kaposi sarcoma and a subset of angiosarcomas. Studies indicate that D2-40 may be a very specific marker for malignant mesothelioma.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.