Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D2887 Calibration Mix, Restek™
SDP

Catalog No. 06801306 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-801-306 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-801-306 Supplier Restek Corporation Supplier No. 31222
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Made with pure, highly characterized neat material and are prepared using NIST-traceable balances and weights.
  • Each ampul is supplied with a data sheet indicating the exact concentration and a sample chromatogram.
  • (C6) n-Hexane (110-54-3), 6% w/w
  • (C7) n-Heptane (142-82-5), 6% w/w
  • (C8) n-Octane (111-65-9), 8% w/w
  • (C9) n-Nonane (111-84-2), 8% w/w
  • (C10) n-Decane (124-18-5), 12% w/w
  • (C11) n-Undecane (1120-21-4), 12% w/w
  • (C12) n-Dodecane (112-40-3), 12% w/w
  • (C14) n-Tetradecane (629-59-4), 12% w/w
  • (C16) n-Hexadecane (544-76-3), 10% w/w
  • (C18) n-Octadecane (593-45-3), 5% w/w
  • (C20) n-Eicosane (112-95-8), 2% w/w
  • (C24) n-Tetracosane (646-31-1), 2% w/w
  • (C28) n-Octacosane (630-02-4), 1% w/w
  • (C32) n-Dotriacontane (544-85-4), 1% w/w
  • (C36) n-Hexatriacontane (630-06-8), 1% w/w
  • (C40) n-Tetracontane (4181-95-7), 1% w/w
  • (C44) n-Tetratetracontane (7098-22-8), 1% w/w

Specifications

Packaging Ampule
Explosion Limit Upper Limit: 6.5% Lower Limit: 0.5%
Ignition Point 202°C
Odor Mild
Flash Point -8°C
Physical Form Liquid
UN Number 1993
Solubility Negligible
Type Multi-component Standard
Quantity 1 mL
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects
Precautionary Statement IF SWALLOWED: Immediately call a POISON CENTER/doctor/.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
ShelfLife 6 months
DOTInformation Class 3: Packing Group II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.