Learn More
Tecan D4+ Dispensehead Cassettes, 4 Well, 1 nL to 10 µL, 20 Cassettes Per Box
Supplier:  Tecan 30097371
Encompass
For use with D300e Digital Dispenser. For non contact dispensing of DMSO and aqueous solutions with a surfactant from 1 nL to 10 L. 20 cassettes/sales unit. With the Tecan D300e Digital Dispenser, you can automatically dispense any volume into any well of any assay plate, quickly and easily. Simply add your liquid tothe Dispensehead and let the instrument do the rest.
Catalog No. NC1202481
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.