Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D609 Potassium Salt 98%, Thermo Scientific™

Catalog No. AC328220010
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328220010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328220010 Supplier Thermo Scientific Chemicals Supplier No. 328220010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83373-60-8
Molecular Formula C11H18KOS2
Molecular Weight (g/mol) 269.482
InChI Key GKKNRBFQLIBXRD-UHFFFAOYSA-N
Synonym carbonodithioic acid o-octahydro-4,7-methano-1h-inden-5-yl ester potassium salt
PubChem CID 131673817
SMILES [HH].C1CC2C(C1)C3CC2CC3OC(=S)S.[K]

Specifications

Color White
Infrared Spectrum Authentic
Quantity 1 mg
Formula Weight 266.5
Percent Purity ≥97.5%
Physical Form Solid
Chemical Name or Material D609 potassium salt, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.