Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501734786 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4786 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4786 Supplier Proteintech Group Inc Supplier No. 252061AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAAM2 is a recently identified planar cell polarity (PCP) signaling molecule implicated in the regulation of cellular polarity, convergent extension, and invasion. DAAM2 belongs to the formin homology family and contains a DAD (diaphanous autoregulatory) domain, an FH1 (formin homology 1) domain, an FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/ formin homology 3) domain. The DAAM2 protein is known to be involved in cell divison. Though the exact function of DAAM2 is yet unknown, reports suggest that it is closely related to DAAM1 and hence along with DAAM1 it may also be required in Wnt/Fz signaling and activation of Rho and in regulating cytoskeleton architecture. DAAM2 is expressed in most of tissues but the expression level ishigher in brain.

Specifications

Antigen DAAM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DAAM2
Gene Accession No. Q80U19, Q86T65
Gene Alias DAAM2, KIAA0381
Gene Symbols Daam2
Host Species Rabbit
Immunogen DAAM2 Fusion Protein Ag18955
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23500, 316201, 76441
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.