Learn More
Abcam DAB Enhancer, Liquid, 10 mL
Description
- Application: Use as a DAB signal enhancer in conjunction with peroxidase-based immunostaining systems.
- Form: Liquid.
- Storage: Store at ambient temperature.
- Shipping: Ambient (cannot ship with ice).
- Constituents: 100% 3,3'-Diaminobenzidine.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 3,3'-Diaminobenzidine |
| Substrate Properties | Chemical Substrate |
| Substrate | DAB |
| Reactivity | Peroxidase |
| Shipping Condition | Ambient |
| Kit Contents | DAB Enhancer, 10 mL |
| Format | Bottle |
| Buffer | 100% 3,3'-Diaminobenzidine |
| Content And Storage | Store at ambient temperature. |
| Form | Liquid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.