Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dab1 Polyclonal Antibody, Rockland™
SDP

Catalog No. p-7191096
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0050 25 μL
50-199-0049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0050 Supplier Rockland Immunochemicals Supplier No. 100401225S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is directed against Dab1 from mouse. Cross-reactivity with other species has not been determined. No reaction occurs with human or mouse Dab2.

The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.

Specifications

Antigen Dab1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 75 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene DAB1
Gene Accession No. P97318
Gene Alias AI956902; C630028C02Rik; Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Dab, reelin signal transducer, homolog 1 (Drosophila); DAB1; DAB1, reelin adaptor protein; disabled 1; Disabled homolog 1; RP23-356E21.1; scm; scr; scrambler; yot; yotari
Gene Symbols DAB1
Host Species Rabbit
Immunogen This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide corresponding to the C-terminal region of murine Dab1 at amino acids 400-555.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13131
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.