Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAB2IP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7210543 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4098 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4098 Supplier Proteintech Group Inc Supplier No. 235821AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.

Specifications

Antigen DAB2IP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DAB2IP
Gene Accession No. Q3UHC7, Q5VWQ8
Gene Alias AF9Q34, AIP1, ASK interacting protein 1, DAB2 interacting protein, DAB2 interaction protein, DAB2IP, DIP1/2, KIAA1743
Gene Symbols Dab2ip
Host Species Rabbit
Immunogen DAB2IP Fusion Protein Ag20308
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153090, 69601
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.