Learn More
Cayman Chemical DabcylKSARSCoV2 ReplIcase 10mg
SDP
Supplier:  Cayman Chemical 3740910 MG
Encompass_Preferred
A fluorogenic substrate for SARS-CoV and SARS-CoV-2 Mpro; upon enzymatic cleavage by SARS-CoV or SARS-CoV-2 Mpro at the Gln-Ser site, EDANS is separated from the Dabcyl quencher and its fluorescence can be used to quantify SARS-CoV or SARS-CoV-2 Mpro activity
Catalog No. 50-291-7780