Learn More
Enzo Life Sciences Dacomitinib (free base) (50mg). CAS: 1110813-31-4
SDP
Supplier:  Enzo Life Sciences ENZCHM2210050
Encompass_Preferred
Dacomitinib is a potent, highly selective, second-generation small-molecule inhibitor of the pan-epidermal growth factor receptor (EGFR) family of tyrosine kinases (ErbB family) with potential antineoplastic activity. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3586
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.