Description
Dade™ Innovin™ Reagent is prepared from purified recombinant human tissue factor produced in E. coli combined with synthetic phospholipids, calcium, buffers, and stabilizers. The reagent contains minimal residual clotting factors, such as prothrombin and factors VII or X, for clear definition of factor deficiencies and steep factor assay curves. It is highly sensitive to extrinsic factor deficiencies and oral anticoagulant-treated patient plasma samples.
The sensitivity of the Dade™ Innovin™ Reagent is very similar to the WHO human brain reference thromboplastin. It is insensitive to therapeutic levels of heparin, which, in combination with its high sensitivity to coagulation factors, makes Dade™ Innovin™ ideal for monitoring oral anticoagulant therapy and differentiating abnormal plasmas, even in the mildly pathological range.
Specifications
Specifications
| Type | Hemostasis Reagent |
| For Use With (Application) | Prothrombin Time (PT) Testing |
| Quantity | 10 x 10 mL |