Learn More
Siemens Healthineers Dade™ PFA-100™ Coagulation Trigger Solution
Catalog No. 23044643 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 11 mL
Catalog No. Quantity
23-044-643 3 x 11 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-643 Supplier Siemens Healthineers Supplier No. 10445701
Add to Cart
Add to Cart
This item is not returnable. View return policy
To aid in the detection of platelet dysfunction in citrated human whole blood.
Dade™ PFA Trigger Solution is an isotonic buffer solution used for triggering the membrane for cartridges for the PFA Systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Coagulation Testing
For Use With (Equipment) PFA-100™ Systems
Quantity 3 x 11 mL
WARNING: Cancer - www.P65Warnings.ca.gov