Learn More
Abcam DAF-2 DA, Fluorescent Nitric Oxide Probe, 1 mg
Description
- Purity: >98%
- Molecular Weight: 446.41 g/mol
- CAS Number: 205391-02-2
- Description: Cell-permeable fluorescent nitric oxide (NO) indicator; deacetylated by intracellular esterases to DAF-2 (ab145269), which reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T) (ab145473)
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Nitric Oxide |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Substrate | Nitric oxide |
| Buffer | 100% DAF-2 DA |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Formulation | 100% DAF-2 DA |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.