Learn More
Cayman Chemical DAF-2 diacetate, 205391-02-2, 250ug
SDP
Supplier:  Cayman Chemical 85165250
Encompass_Preferred
Molecular Formula C24H18N2O7, Purity ≥95%, Formula Weight 446.4, Synonyms: 2-(3,6-diacetyloxy-4,5-diamino-9H-xanthen-9-yl)-benzoic acid, 4,5-Diaminofluorescein diacetate; DAF-2 diacetate is a sensitive fluorescent indicator for the detection and bioimaging of NO.{6360,6632} It is a cell-permeable derivative of DAF-2. Upon entry into the cell, DAF-2 diacetate is transformed into the less cell-permeable DAF-2 by cellular esterases thus preventing loss of signal due to diffusion of the molecule from the cell. In the presence of oxygen, DAF-2 reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T). Fluorescence is monitored using excitation and emission wavelengths of 485 and 538 nm, respectively.{6632} At neutral pH the detection limit for NO is 2-5 nM. DAF-2 diacetate can be utilized in cells which produce small amounts of NO, such as endothelial cells, as well as in cells which generate large amount of NO, such as macrophages.{6360,6632}.
Catalog No. NC0190930
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.