Learn More
Abcam DAF-FM DA, Nitric Oxide Quantification Reagent, >98% Purity, 1 mg
Description
- Molecular Weight: 496.42 g/mol
- Purity: >98%
- Application: Cell permeable reagent for quantification of low concentrations of nitric oxide in solution
- Mechanism: Essentially nonfluorescent until hydrolyzed to DAF-FM by intracellular esterases and reacts with NO to form a fluorescent benzotriazole
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Nitric Oxide |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Substrate | Nitric oxide |
| Buffer | 100% DAF-FM DA |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection, In-Solution Detection |
| Form | Solid |
| Kit Contents | 1 mg DAF-FM DA |
| Detection Method | Fluorescence |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.