Learn More
Abcam DAF-FM, Nitric Oxide Fluorescent Probe, >98% Purity, 1 mg
Description
- Highly sensitive and photo-stable fluorescent probe for the quantification of low concentrations of nitric oxide (NO)
- More sensitive to NO than DAF-2 (ab145269)
- More photo-stable and less pH sensitive than DAF-2
- Does not require activation by cytosolic enzymes
- Essentially nonfluorescent until reaction with NO forms a fluorescent benzotriazole
- Purity: >98%
- Molecular Weight: 412.34 g/mol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Substrate | Nitric oxide |
| Target Molecule | Nitric Oxide |
| Shipping Condition | Ambient |
| Reactivity | Nitric oxide |
| Label or Dye | Fluorescent Probe |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Form | Solid |
| Detection Method | Fluorescence |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.