Learn More
Abcam DAF-FM, Nitric Oxide Fluorescent Probe, >98% Purity, 5 mg
Description
- Highly sensitive and photo-stable fluorescent probe for the quantification of low concentrations of nitric oxide (NO).
- More sensitive to nitric oxide than DAF-2 (ab145269).
- More photo-stable and less pH-sensitive than DAF-2.
- Does not require activation by cytosolic enzymes.
- Essentially nonfluorescent until reaction with nitric oxide to form a fluorescent benzotriazole.
- Molecular weight: 412.34 g/mol.
- Purity: >98%.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Nitric Oxide |
| Label or Dye | Fluorescent Probe |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | Nitric oxide |
| Format | Vial |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.