Learn More
Enzo Life Sciences Diacylglycerol Kinase Inhibitor II (R59 949) (25mg). CAS: 120166-69-0
SDP
Supplier:  Enzo Life Sciences ALX430028M025
Encompass_Preferred
Inhibitor of diacylglycerol kinase in intact cells (IC50=0.12µM) and in isolated membranes (IC50=0.3µM). More potent analog of R59 022 (ALX-430-027). Useful for elucidating roles of protein kinase C (PKC). Alternative name: 3-[2-[4-[bis(4-Fluorophenyl)methylene]-1-piperidinyl]ethyl]-2,3-dihydro-2-thioxo-4(1H)-quinazolinone. Purity: ≥98% (HPLC). Solubility: Soluble in dichloromethane, ethyl acetate, tetrahydro-furan, DMSO (33mg/ml) or dimethyl formamide; almost insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9159
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.