Learn More
Enzo Life Sciences Daidzin (10mg). CAS: 552-66-9
SDP
Supplier:  Enzo Life Sciences ALX350248M010
Encompass_Preferred
Glucoside of the isoflavone daidzein found in soy beans. Alternative name: Daidzein-7-O-glucoside. Purity: ≥98%. Appearance: White to beige powder. Solubility: Soluble in DMSO or dilute aqueous base; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8803
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.