Learn More
Electron Microscopy Sciences Dakin-Fully Frosted Slide
SDP
Supplier:  Electron Microscopy Sciences 7187610
Encompass_Preferred
Special glass slides measuring 1x3" (25x75 mm) and 1 mm thick. A fully frosted sandblasted surface on the same side as the superfrosted end.
Catalog No. 50-949-742
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.