Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dalapon, Restek
SDP

Catalog No. p-2714194 Shop All Restek Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Concentration or Composition (by Analyte or Components):
1000 μg/mL in Methanol
2000 μg/mL in Methanol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration or Composition (by Analyte or Components)
06-719-878 1 mL 1000 μg/mL in Methanol
06-719-704 1 mL 2000 μg/mL in Methanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-719-878 Supplier Restek Corporation Supplier No. 32253
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-99-0
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.963
InChI Key NDUPDOJHUQKPAG-UHFFFAOYSA-N
PubChem CID 6418
ChEBI CHEBI:81859
IUPAC Name 2,2-dichloropropanoic acid
SMILES CC(C(=O)O)(Cl)Cl

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000 μg/mL in Methanol
Chemical Name or Material Dalapon, Herbicide Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.