Learn More
TARGETMOL CHEMICALS INC DIM-C-PPHOH 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4400
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DIM-C-pPhOH (CDIM8) is a nur77 (NR4A1) antagonistinhibits TGF-(beta) induced cell migration of breast cancer cell linespromotes ROS/endoplasmic reticulum stress and proapoptotic pathways in pancreatic cancer cell linesmimics effects of Nur77 RNAi silencing. purity: 99%
Catalog No. 50-257-9887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.