Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DAN/NBL1 Antibody Pair [HRP]
SDP

Catalog No. NB009979 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009979 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009979 Supplier Novus Biologicals™ Supplier No. NBP27947015PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 46.9 pg/mL
Assay Sensitivity 46.88-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human DAN/NBL1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias DAN domain family member 1, MGC8972, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN
Gene ID (Entrez) 4681
Gene Symbol NBL1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 110 ng of recombinant DAN/NBL1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-DAN/NBL1 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL mouse anti-DAN/NBL1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cell Cycle and Replication
    Storage Requirements Storage is content dependent.
    Target DAN/NBL1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 3 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.