Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAND5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583970 Supplier Invitrogen™ Supplier No. PA583970
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMCKAVPFVQ VFSRPGCSAI RLRNHLCFGH CSSLYIPGSD PTPLVLCNSC MPARKRWAPV VLWCLTGSSA SRRRVKISTM LIEGCHCSPK A Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 65%.

This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to bind Nodal and to inhibit the Nodal signaling pathway which patterns left/right body asymmetry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAND5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DAND5
Gene Accession No. Q8N907
Gene Alias CER2; CER2CRL2; cerberus 2; Cerberus like-2; cerberus-like 2; cerberus-like protein 2; Cerl2; Cerl-2; Cerr2; CKTSF1B3; CKTSF1B3MGC126849; coco; CRL2; Cysteine knot superfamily 1, BMP antagonist 3; DAN domain BMP antagonist family member 5; DAN domain family member 5; DAN domain family member 5, BMP antagonist; DAN domain family, member 5; Dand5; DANTE; Dte; FLJ38607; GREM3; GREM3cerberus 2; gremlin-3; MGC126849; SP1
Gene Symbols DAND5
Host Species Rabbit
Immunogen Recombinant protein fragment of Human DAND5 (Product #RP-99178)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.