Learn More
Advanced Materials Technology DANDYVICE WITH HOLDER
Supplier:  Advanced Materials Technology 94900009
Encompass_Preferred
The dandyVice is a tool that assists in the process of the routine filtration in a laboratory. This tool is ideal for routine sample filtration accommodating syringe sizes of 1 ml – 20 ml and associated filter discs. The concept is aimed at reducing pain and fatigue of the hand and fingers.
Catalog No. 50-255-5993
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.