Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Danger: High Voltage

Catalog No. 19804569 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
3.5 x 5 in.
5 x 7 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-804-569 3.5 x 5 in.
19-804-570 5 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-804-569 Supplier Accuform Signs Supplier No. LELC248XVE
Add to Cart
Add to Cart

Colorful, compact safety signs

  • Legend: DANGER: HIGH VOLTAGE
  • Available in a variety of sizes and materials
  • Color: White, black, red
  • Language: English

Model LELC036VSP

  • Legend: DANGER. HIGH VOLTAGE INSIDE

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Finish 2 mil High Gloss UV Polyester over laminate film
Material Vinyl
Quantity 1 Ea.
Description Dura Vinyl
For Use With (Application) Provides proper protection where head protection is required for outdoor usage
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Thickness 6 mil
Width (English) 5 in.
Width (Metric) 12.7 cm
Temperature Range (Metric) -28°C to 80°C
Temperature Range (English) -20°F to 176°F
Languages English
Dimensions (L x W) 3.5 x 5 in.
Displayed Text DANGER High Voltage
Product Type Electrical Hazard Sign
Color Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.