Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco DANi Teen and Young Adult Advanced Patient Care Simulator - Medium
SDPSCI_ED

Catalog No. S97916 Shop All Simulaids, Inc. Products
Change view
Click to view available options
Color:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S97916 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S97916 Supplier Nasco Supplier No. LF04504M
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

DANi is an advanced training solution designed to support training on the fundamentals of nursing and EMS procedures for issues relating to drug abuse, sexual assault, mental health, or self-harm.

DANi accurately reproduces scenarios and challenging dynamics for learners to administer critical care for this important patient demographic. Learners can practice on a comprehensive and complete simulation platform including patient monitoring capability and auscultation (heart and lung sounds).

A realistic, hands-on learning experience ideally suited for classroom, in-situ, or hospital instruction.

Basic Life Support (BLS) and Advanced Cardiac Life Support (ACLS)

CPR with quality performance metrics, carotid pulse, intubation

Trauma Simulation

  • Physical assessment
  • Airway, breathing, circulation
  • Body trauma scan
  • Wound care
  • Self-harm/drug use moulage
  • Asymmetric pupils

Realistic Features

  • Pupils
  • Airway
  • Acne
  • Smooth skin
  • Tattoo sleeve
  • Piercings and earrings
  • Dyed and black wigs
  • Foot wounds

Nursing Care

  • Grooming
  • Daily living assistance
  • Eye/ear/hair care
  • Catheter
  • Injection
  • Oxygen
  • Blood pressure
  • Pericare
  • Ostomy care
  • Urinary catheter care
  • Oxygen administration

Female and Male Genitalia

  • Includes female sexual assault genitalia

Patient Positioning

  • Superior range of motion, seating, laying, finger and toe manipulation

Respiratory Support

  • Manual ventilation with chest rise
  • Intubation
  • Ventilation
  • Tracheostomy care
  • Suctioning

Auscultation

  • 6 anterior heart sites with 12 heart conditions
  • 5 anterior, 6 upper posterior, 4 lower posterior
  • 2 mid-axillary lung sites with 12 lung conditions

Advanced Skills

  • Gastrostomy
  • Nasogastric procedures
  • Airway/trach suctioning
  • Enema
  • Pap smear
  • Pelvic exam
  • Intravenous injections

SKILLQUBE™ Patient Monitoring/Defibrillator

  • ECG monitor (with additional patient vitals and monitored signals: ABP, SpO2, etCO2)
  • Pacemaker
  • Defibrillator
  • AED
  • Ventilator (generic)
  • Instructor controlled scenario editor with library
  • Auscultation, glucometer, thermometer, checklists, operating procedures
  • Realistic laboratory values
  • Optional CPR feedback with real-time evaluation (add-on)

Specifications

Product Type Teen and Young Adult Patient Care Simulator
For Use With (Application) Simulating Patient Care
Color Medium
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.