Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco DANi Teen and Young Adult Advanced Patient Care Trainer - Light
SDPSCI_ED

Catalog No. S97911 Shop All Simulaids, Inc. Products
Change view
Click to view available options
Color:
Light
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S97911 Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S97911 Supplier Nasco Supplier No. LF04500
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

DANi is an advanced training solution designed to support training on the fundamentals of nursing and EMS procedures for issues relating to drug abuse, sexual assault, mental health, or self-harm.

DANi accurately reproduces scenarios and challenging dynamics for learners to administer critical care for this important patient demographic. Learners can also practice general patient care, patient positioning, bandaging and wound dressing, oxygen administration.

A realistic, hands-on learning experience ideally suited for classroom, in-situ, or hospital instruction.

Trauma Simulation

  • Physical assessment
  • Airway, breathing, circulation
  • Body trauma scan
  • Wound care
  • Self-harm/drug use moulage
  • Asymmetric pupils

Realistic Features

  • Pupils
  • Acne
  • Smooth skin
  • Tattoo sleeve
  • Piercings and earrings
  • Dyed, brown and black wigs
  • Foot wounds

Nursing Care

  • Grooming
  • Daily living assistance
  • Eye/ear/hair care
  • Oxygen
  • Pericare
  • Ostomy care
  • Urinary catheter care
  • Oxygen administration

Female and Male Genitalia

  • Includes female sexual assault genitalia

Patient Positioning

  • Superior range of motion, seating, laying, finger and toe manipulation

Specifications

Product Type Teen and Young Adult Patient Care Trainer
For Use With (Application) Simulating Patient Care
Color Light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.