Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansyl chloride, 96%

Catalog No. p-7021480
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1382803 1 g
AAA1382806 5 g
AAA1382814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1382803 Supplier Thermo Scientific Chemicals Supplier No. A1382803
Add to Cart

CAS: 605-65-2 | C12H12ClNO2S | 269.74 g/mol

Dansyl chloride is a fluorogenic reagent used for protein, N-terminal amino acid and peptide detection by reverse phase high performance liquid chromatography (HPLC). It is also used to determine the degree of labeling in chymotrypsin and ovalbumin. It acts as a precursor for the synthesis of other derivatives like dansyl amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dansyl chloride is a fluorogenic reagent used for protein, N-terminal amino acid and peptide detection by reverse phase high performance liquid chromatography (HPLC). It is also used to determine the degree of labeling in chymotrypsin and ovalbumin. It acts as a precursor for the synthesis of other derivatives like dansyl amide.

Solubility
Soluble in dimethyl fluoride, acetone, chloroform, pyridine, benzene and dioxane. Insoluble in water.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-65-2
Molecular Formula C12H12ClNO2S
Molecular Weight (g/mol) 269.74
MDL Number MFCD00003985
InChI Key XPDXVDYUQZHFPV-UHFFFAOYSA-N
Synonym dansyl chloride, 5-dimethylamino naphthalene-1-sulfonyl chloride, dns chloride, dansyl, 5-dimethylamino-1-naphthalenesulfonyl chloride, 1-naphthalenesulfonyl chloride, 5-dimethylamino, 1-dimethylaminonaphthalene-5-sulfonyl chloride, 1-chlorosulfonyl-5-dimethylaminonaphthalene, unii-qmu9166tj4, 5-dimethylaminonaphthalene-1-sulphonyl chloride
PubChem CID 11801
ChEBI CHEBI:51907
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
SMILES CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O

Specifications

Melting Point 69°C to 75°C
Odor Odorless
Quantity 1 g
UN Number UN3261
Beilstein 2217205
Sensitivity Moisture sensitive
Merck Index 14,2814
Solubility Information Soluble in dimethyl fluoride,acetone,chloroform,pyridine,benzene and dioxane. Insoluble in water.
Formula Weight 269.75
Percent Purity 96%
Chemical Name or Material Dansyl chloride
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-092-6
RTECSNumber QK3688000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.