Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dansyl Chloride, Fluorogenic Amine Reactive Reagent, Solid, 1 g

Catalog No. 502704655
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4655 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4655 Supplier Abcam Supplier No. AB1463971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic primary amine reactive reagent for N-terminal derivatization of amino acids and peptides.

  • Molecular Weight: 269.75 g/mol
  • Purity: >95%
  • Form: Solid
  • Applications: Fluorogenic primary amine reactive reagent for N-terminal derivatization of amino acids and peptides, and for polyamine detection by reverse phase HPLC and other analytical procedures
  • Fluorescence: λex 337 nm; λem 492 nm in chloroform (after derivatization with hexylamine)
  • Composition: 100% Dansyl chloride
  • CAS Number: 605-65-2

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Primary Amine
Substrate Properties Chemical Substrate
Substrate Primary amine
Shipping Condition Ambient
For Use With (Equipment) HPLC
Product Type Protein Labeling Reagent
Buffer 100% Dansyl chloride
Detection Method Fluorescence
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Dansyl chloride
Quantity 1 g
For Use With (Application) N-terminal derivatization of amino acids and peptides, polyamine detection by reverse phase HPLC, analytical procedures
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.