Learn More
Abcam Dansyl Chloride, Fluorogenic Primary Amine Reactive Reagent, 5 g
Description
- Molecular Weight: 269.75 g/mol
- Purity: >95%
- Physical Form: Solid
- Applications: N-terminal derivatization of amino acids and peptides; polyamine detection by reverse phase HPLC and other analytical procedures
- Fluorescence: λex 337 nm; λem 492 nm in chloroform (after derivatization with hexylamine)
- Composition: 100% Dansyl chloride
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Primary Amines |
| Substrate | Primary amine |
| For Use With (Equipment) | HPLC |
| Product Type | Protein Labeling Reagent |
| Form | Solid |
| Detection Method | Fluorescence |
| Buffer | 100% Dansyl chloride |
| Content And Storage | Store at -20°C. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.