Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAOA Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734493 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4493 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4493 Supplier Proteintech Group Inc Supplier No. 246921AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This gene encodes a protein that is an activator of the FAD-dependent enzyme D-amino acid oxidase, which degrades the gliotransmitter D-serine, a potent activator of N-methyl-D-aspartate (NMDA) type glutamate receptors. Polymorphisms in this gene have been implicated in susceptibility to schizophrenia and bipolar affective disorder, possibly due to decreased levels of D-serine and decreased NMDA receptor functioning. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen DAOA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DAOA
Gene Accession No. P59103
Gene Alias D amino acid oxidase activator, DAOA, G72, LG72, Protein G72, SG72
Gene Symbols DAOA
Host Species Rabbit
Immunogen DAOA Fusion Protein Ag19143
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 267012
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.