Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP5 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7144552
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2514 10 μL
50-157-2513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2514 Supplier Bethyl Laboratories, Inc Supplier No. IHC00673T
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, G

Eukaryotic translation initiation factor 4 gamma 2 (elF4G or DAP5) plays a role in the switch from cap-dependent to IRES-mediated translation during mitosis, apoptosis, and viral infections. eIF4G2 is a ubiquitously expressed member of the translation initiation factor eIF4G family that lacks the EIF4E binding site.

Specifications

Antigen DAP5
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EIF4G2
Gene Accession No. P78344
Gene Alias AAG1, DAP5, FLJ41344, NAT1, P97
Gene Symbols EIF4G2
Host Species Rabbit
Immunogen Between 125 and 175
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1982
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.