Learn More
Invitrogen™ DAP5 Polyclonal Antibody

Description
Positive test controls include: HeLa, 293T, Jurkat, LO2, Mouse brain. Immunogen sequence: PKTINQIRQD AVKDLGVFIP APMAQGMRSD FFLEGPFMPP RMKMDRDPLG GLADMFGQMP GSGIGTGPGV IQDRFSPTMG RHRSNQLFNG HGGHIMPPTQ SQFGEMGGKF MKSQGLSQLY HNQSQGLLSQ LQGQSKDMPP RFSKKGQLNA DEISLRPAQS FLMNKNQVPK L.
Specifications
Specifications
| Antigen | DAP5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Eif4g2 |
| Gene Accession No. | P78344, Q62448 |
| Gene Alias | AA589388; AAG1; aging-associated protein 1; DAP5; DAP-5; Death-associated protein 5; E130105L11Rik; eIF4G 2; eIF-4G 2; EIF4G2; eif4g2.L; eif4g2.S; eIF-4-gamma 2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4 gamma, 2 L homeolog; eukaryotic translation initiation factor 4 gamma, 2 S homeolog; eukaryotic translation initiation factor 4 gamma, 2 S homeolog; eukaryotic translation initiation factor 4, gamma 2; eukaryotic translation initiation factor 4G-like 1; FLJ41344; NAT1; Natm1; Novel APOBEC-1 target 1; OK/SW-cl.75; p97; translation repressor NAT1; XELAEV_18024574mg |
| Gene Symbols | Eif4g2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.