Learn More
Medchemexpress LLC HY-10450A 5mg Medchemexpress, Dapagliflozin ((2S)-1,2-propanediol, hydrate) CAS:960404-48-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10450A5MG
Encompass_Preferred
Medchemexpress, HY-10450A 5mg Dapagliflozin ((2S)-1,2-propanediol, hydrate) CAS:960404-48-2 Dapagliflozin ((2S)-1,2-propanediol, hydrate) is the S-enantiomer of Dapagliflozin 1,2-propanediol, hydrate. Dapagliflozin ((2S)-1,2-propanediol, hydrate), a new type of drug used to treat diabetes mellitus (DM), is a competitive sodium/glucose cotransporter 2 (SGLT2) inhibitor, which results in excretion of glucose into the urine.Dapagliflozin ((2S)-1,2-propanediol, hydrate) induces HIF1 expression and attenuates renal IR injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8224
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.