Learn More
Apexbio Technology LLC BV6, 100mg. Cas: 1001600-56-1 MFCD: MFCD28168021. Selective inhibitor of IAP proteins
SDP
Supplier:  Apexbio Technology LLC B4653100
Encompass_Preferred
BV6 is a selective inhibitor of IAP family with IC50 value of 7.2 uM when tested with H460 cells. 1001600-56-1. MFCD28168021
Catalog No. 50-101-5395
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.