Learn More
BIOGEMS INTERNATIONAL INC DAPI (Hydrochloride) 5mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 28790385MG
Encompass_Preferred
DAPI is a cell-permeable fluorescent stain that binds to the A-T rich regions of DNA and less strongly to RNA as well. It has an absorption maximum at the 358 nm (ultraviolet) wavelength and an emission maximum at 461 nm (blue) when bound to DNA.
Catalog No. 50-152-9990
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.