Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daple Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563888
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3888 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3888 Supplier Bethyl Laboratories, Inc Supplier No. A302951AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a ubiquitously expressed coiled-coil domain-containing protein that interacts with the disheveled protein and is a negative regulator of the Wnt signaling pathway. The protein encoded by this gene has a PDZ-domain binding motif in its C-terminus with which it interacts with the disheveled protein. Disheveled is a scaffold protein involved in the regulation of the Wnt signaling pathway. The Wnt signaling pathway plays an important role in embryonic development, tissue maintece, and cancer progression. Mutations in this gene cause autosomal recessive, primary non-syndromic congenital hydrocephalus; a condition characterized by excessive accumulation of cerebrospinal fluid in the ventricles of the brain.

Specifications

Antigen Daple
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CCDC88C
Gene Accession No. Q9P219
Gene Alias Coiled-coil domain-containing protein 88C, DAPLE, Dvl-associating protein with a high frequency of leucine residues, hDaple, HkRP2, hook-related protein 2, KIAA1509, SCA40, spinocerebellar ataxia 40
Gene Symbols CCDC88C
Host Species Rabbit
Immunogen Between 1450 and 1500
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440193
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.