Learn More
Medchemexpress LLC HY-B0304AS 5mg Medchemexpress, Dapoxetine-D7 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0304AS5MG
Encompass_Preferred
Medchemexpress, HY-B0304AS 5mg Dapoxetine-D7 (hydrochloride) CAS: Dapoxetine-D7 (LY-210448-D7) hydrochloride is the deuterium labeled Dapoxetine hydrochloride. Dapoxetine hydrochloride is a short-acting selective serotonin reuptake inhibitor (SSRI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2457