Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dapoxetine hydrochloride, Tocris Bioscience™

Catalog No. 435810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-581-0 10 mg
43-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-581-0 Supplier R&D Systems Supplier No. 4358/10
This item is not returnable. View return policy

Selective serotonin reuptake inhibitor

Selective serotonin reuptake inhibitor (SSRI).

Chemical Identifiers

CAS 129938-20-1
Molecular Formula C21H24ClNO
Molecular Weight (g/mol) 341.879
InChI Key IHWDIQRWYNMKFM-BDQAORGHSA-N
Synonym dapoxetine hydrochloride, dapoxetine hcl, priligy, ly 210448 hydrochloride, unii-u4oht63mri, u4oht63mri, ly-210448 hydrochloride, dapoxetine hydrochloride usan, s-n,n-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine hydrochloride
PubChem CID 71352
IUPAC Name (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine;hydrochloride
SMILES CN(C)C(CCOC1=CC=CC2=CC=CC=C21)C3=CC=CC=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 341.87
Percent Purity >99%
Chemical Name or Material Dapoxetine hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.