Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583804 Supplier Invitrogen™ Supplier No. PA583804
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRNELKYFKD QMSPEPIRIL DLTECSAVQF DYSQERVNCF CLVFPFRTFY LCAKTGVEAD EWIKILRWKL SQIRKQLNQG Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 96%.

May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAPP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DAPP1
Gene Accession No. Q9UN19
Gene Alias b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1
Gene Symbols DAPP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DAPP1. Recombinant protein control fragment (Product #RP-96506).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27071
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.