Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dapsone, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602020 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-020 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-020 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D134725KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

D1347, 80-08-0, C12H12N2O2S

Spectrum™ Chemical Dapsone, USP belongs to a class of drugs known as sulfones that is used to treat a certain type of skin disorder (dermatitis herpetiformis). All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 80-08-0
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.30
InChI Key MQJKPEGWNLWLTK-UHFFFAOYSA-N
IUPAC Name 4-(4-aminobenzenesulfonyl)aniline
SMILES NC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(N)C=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.015
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.