Learn More
AdipoGen DAPT (25 mg)
SDP
Supplier:  AdipoGen AGCR10016M025
Encompass_Preferred
DAPT (25 mg), AdipoGen Life Sciences. Chemical. CAS: 208255-80-5. Formula: C23H26F2N2O4. MW: 432.5. Cell permeable gamma-secretase inhibitor (IC50 = 115 nM for total beta-amyloid, IC50 = 200 nM for beta-amyloid 1-42). Reduces Abeta levels in vivo. Blocks the proteolytic processing of neurotrophin receptor alike death domain protein (NRADD). Does not inhibit persenilinase. Notch processing inhibitor. Enhances neuronal differentiation independent of sonic hedgehog (Shh) signaling. CDK5 activity inhibitor. Apoptosis enhancer.
Catalog No. 50-149-0052
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.