Learn More
Apexbio Technology LLC DAPT (GSI-IX), 50mg. Cas: 208255-80-5 MFCD: MFCD04974585. γ-secretase inhibitor,potent and specific
SDP
Supplier:  Apexbio Technology LLC A820050
Encompass_Preferred
DAPT, N-[N-(3,5-Difluorophenacetyl)-L-alanyl]-S-phenylglycine t-butyl Ester, is a potent and specific inhibitor of -secretase, a multimeric MEM protein complex that catalyzes proteolytic cleavage of amyloid precursor protein (APP) resulting in the accumulation of amyloi- (A) peptides. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5074
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.