Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Daptomycin
GREENER_CHOICE

Catalog No. p-4817510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-171-0 10 mg
39-175-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-171-0 Supplier Tocris Bioscience™ Supplier No. 3917/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antibiotic; inhibits protein, DNA and RNA synthesis in gram-positive bacteria

Daptomycin is a lipopeptide, calcium-dependent antibiotic. Exhibits potent bacteriocidal activity against most gram-positive bacteria in vitro and in vivo, including antibiotic-resistant strains such as MRSA and VRE. Disrupts plasma membrane function; activity results in membrane depolarization leading to inhibition of protein, DNA and RNA synthesis.

Chemical Identifiers

CAS 103060-53-3
Molecular Formula C72H101N17O26

Specifications

Quantity 10 mg
Formula Weight 1620.67
Product Type Daptomycin
Purity >95%
Molecular Weight (g/mol) 1620.693
Chemical Name or Material Daptomycin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.